Yohimbine hydrochloride, 98%
Yohimbine hydrochloride, 98%
Yohimbine hydrochloride, 98%
Acros Organics

Yohimbine hydrochloride, 98%

Yohimbine hydrochloride, CAS # 65-19-0, is an α2-adrenergic receptor antagonist, which blocks the central α2-adrenegic receptors in the brain. | CAS: 65-19-0 | C21H27ClN2O3 | 390.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 141030250
also known as 14103
HSN 29339900
Price (INR)
56,072.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
56,072.00
Each
Chemical Identifiers
CAS65-19-0
IUPAC Namehydrogen methyl (1S,15R,18S,19R,20S)-18-hydroxy-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate chloride
Molecular FormulaC21H27ClN2O3
InChI KeyPIPZGJSEDRMUAW-VJDCAHTMSA-N
SMILES[H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Specific optical rotation+99° to +105° (20°C, 589 nm) (c=1, water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Yohimbine hydrochloride is an indole alkaloid derived from the tree bark
  • It is an antagonist of α2-adrenergic receptors in the brain, reducing the effects of xylazine, which is an α2-adrenergic agonist

Applications

  • Yohimbine hydrochloride blocks the central α2-adrenegic receptors in the brain
  • It can reduce or eradicate effects of xylazine, which is an α2-adrenergic agonist
  • It can have an antidiuretic effect
RUO – Research Use Only

General References:

  1. Zhang, W. and Roomans, G.M. A yohimbine-dependent, UK14,304 induced ion transient in HT29 cells studied by X-ray microanalysis. Scanning microscopy, 1996, 10 (1), 293-299.
  2. Susan, X. Jiang et. al. Reversible inhibition of intracellular calcium influx through NMDA receptors by imidazoline I2receptor antagonists. European Journal of Pharmacology. 2010, 629,(1), 12-19.