EBSS, no calcium, no magnesium, no phenol red
Gibco™

EBSS, no calcium, no magnesium, no phenol red

Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation,자세히 알아보기
Have Questions?
카탈로그 번호수량
14155063500 mL
카탈로그 번호 14155063
제품 가격(KRW)
56,000
온라인 행사
Ends: 30-Jun-2026
62,000
할인액 6,000 (10%)
Each
수량:
500 mL
Customize this product
제품 가격(KRW)
56,000
온라인 행사
Ends: 30-Jun-2026
62,000
할인액 6,000 (10%)
Each
Earle's Balanced Salt Solution (EBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Gibco™ EBSS is formulated without calcium and magnesium for rinsing chelators from the culture before cell dissociation.

This EBSS is manufactured as follows:

WithWithout
• Glucose• Calcium
• Sodium Bicarbonate• Magnesium
• Phenol Red

The complete formulation is available.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ EBSS is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ EBSS product made in our Scotland facility (14155-048). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

This product is not used for in vitro diagnostic purpose in some countries.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
사양
화학물질 이름 또는 재질Earle's Balanced Salt
포함No Sodium Pyruvate
제조 품질cGMP-compliant under the ISO 13485 standard
삼투압270 - 310 mOsm/kg
권장 스토리지Store at room temperature.

Shelf life: 36 months from date of manufacture
멸균Sterile-filtered
농도1 X
용도(애플리케이션)Variety of Cell Culture Applications; Such as Washing Cells Before Dissociation; Transporting Cells or Tissue Samples; Diluting Cells for Counting; and Preparing Reagents
물리적 형태Liquid
제품라인Gibco
수량500 mL
용액 유형Earle's Balanced Salt
pH7.0 - 8.1
Unit SizeEach

자주 묻는 질문(FAQ)

I understand that some media are worse than others for fluorescence imaging. How do I choose?

Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?

Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.