Dulbecco's Phosphate-Buffered Saline (D-PBS) (1X) liquid, SuperCase -"DISCONTINUED" - FAQs
Dulbecco's Phosphate-Buffered Saline (D-PBS) (1X) liquid, SuperCase -"DISCONTINUED" - FAQs
View additional product information for Dulbecco's Phosphate-Buffered Saline (D-PBS) (1X) liquid, SuperCase -"DISCONTINUED" - FAQs (14190318)
3 product FAQs found
What is the specific gravity of DPBS (without calcium and magnesium)?
The specific gravity of this product is approximately 1.006 kg/L.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the overall molarity of DPBS (without calcium and magnesium)?
The overall molarity of this solution is 19.66. This is calculated by taking the sum of the dry weights and dividing it by the sum of the formula weight.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is a precipitate in DPBS (without calcium and magnesium) normal?
No, this is not normal and if there is a precipitate in the product, it cannot be used. If the precipitate will not go back into solution, please contact Technical Support at techsupport@thermofisher.com for a replacement.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.