DPBS, no calcium, no magnesium
Gibco™

DPBS, no calcium, no magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
1419036720 LBag
14190094
also known as 14190-094
500 mLBottle
14190144500 mLBottle
1419025010 x 500 mLBottle
141901361000 mLBottle
141902356 x 1000 mLBottle
141903425 LBag
1419035910 LBag
Catalog number 14190367
Price (USD)
812.65
Online exclusive
874.00
Save 61.35
Each
Quantity:
20 L
Packaging:
Bag
Customize this product
Price (USD)
812.65
Online exclusive
874.00
Save 61.35
Each

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality270 to 300 mOsm/kg
PackagingBag
Recommended StorageStorage conditions: 15°C to 30°C, Shipping conditions: Ambient, Shelf life: 12 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration1X
For Use With (Application)Mammalian Cell Culture
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity20 L
Solution TypeCleaning, Storage, pH Buffer
pH7.0 to 7.3
Unit SizeEach

Frequently asked questions (FAQs)

What is the shelf life of DPBS after it has been opened?

We cannot provide an official expiration date for open bottles as this depends on the customer's use. Once opened, Thermo Fisher Scientific can no longer guarantee the performance of any product. However, if the storage conditions are followed as recommended, the stated expiration date (36 months from the date of manufacture) should apply to opened bottles as DPBS is extremely stable.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the shelf life of DPBS (Cat. No. 14190136)?

DPBS has a shelf life of 36 months from the date of manufacture, when stored as recommended (15-30 degrees C).

Does Diploid Growth Serum-Reduced Medium (SRM) contain L-glutamine?

The basal Diploid Growth Serum-Reduced Medium already contains 6 mM L-glutamine.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do I have to adjust my infection parameters when using Diploid Production Serum-Free Medium (SFM)?

We recommend evaluating performance with Diploid Production Serum-Free Medium using current conditions, however, multiplicity of infection, time of infection, and time of harvest may be different than with conventional media. Closely monitor cells for cytopathic effect and evaluate viral titers at multiple time points.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Can Diploid Growth Serum-Reduced Medium (SRM) be supplemented with different types of serum?

Diploid Growth Serum-Reduced Medium can be supplemented with Fetal Bovine Serum, Newborn Calf Serum, Donor Bovine Serum with Iron, or Bovine Serum.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.