Thermo Scientific™

Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers

Catalog number: 1424442
Thermo Scientific™

Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers

Catalog number: 1424442
For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.
 
Catalog Number
1424442
Unit Size
Each
Wavelength
444 nm
Price (USD)
646.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength444 nm
Excitation/Emission444 nm, HBW 12 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
Price (USD)
314.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeLow Plate Adapter
For Use With (Equipment)Luminoskan™, Fluoroskan™ FL
Unit SizeEach
Catalog Number
1423202
also known as 4232022
Unit Size
Wavelength
320 nm
Price (USD)
672.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength320 nm
Excitation/Emission320 nm, HBW 40 nm
Unit Sizeundefined
Catalog Number
Unit Size
Each
Wavelength
335 nm
Price (USD)
1,644.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength335 nm
Excitation/Emission335 nm, HBW 13 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
340 nm
Price (USD)
1,252.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength340 nm
Excitation/Emission340 nm, HBW 10 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
355 nm
Price (USD)
672.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength355 nm
Excitation/Emission355 nm, HBW 38 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
360 nm
Price (USD)
1,230.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength360 nm
Excitation/Emission360 nm, HBW 14 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
370 nm
Price (USD)
1,292.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength370 nm
Excitation/Emission370 nm, HBW 12 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
380 nm
Price (USD)
1,234.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength380 nm
Excitation/Emission380 nm, HBW 11 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Wavelength
390 nm
Price (USD)
672.00
Your Price:
Availability
***
Full specifications
Quantity
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength390 nm
Excitation/Emission390 nm, HBW 20 nm
Unit SizeEach
Showing 10 of 67Show more
Catalog NumberSpecificationsUnit SizeWavelengthPrice (USD)AvailabilityQuantity
1424442Full specifications
Each444 nm
646.00
Your Price:
***
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength444 nm
Excitation/Emission444 nm, HBW 12 nm
Unit SizeEach
11272700Full specifications
Each
314.00
Your Price:
***
Product TypeLow Plate Adapter
For Use With (Equipment)Luminoskan™, Fluoroskan™ FL
Unit SizeEach
1423202
also known as 4232022
Full specifications
320 nm
672.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength320 nm
Excitation/Emission320 nm, HBW 40 nm
Unit Sizeundefined
1423352Full specifications
Each335 nm
1,644.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength335 nm
Excitation/Emission335 nm, HBW 13 nm
Unit SizeEach
142340210Full specifications
Each340 nm
1,252.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength340 nm
Excitation/Emission340 nm, HBW 10 nm
Unit SizeEach
1423552Full specifications
Each355 nm
672.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength355 nm
Excitation/Emission355 nm, HBW 38 nm
Unit SizeEach
1423602Full specifications
Each360 nm
1,230.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength360 nm
Excitation/Emission360 nm, HBW 14 nm
Unit SizeEach
142370212Full specifications
Each370 nm
1,292.00
Your Price:
***
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength370 nm
Excitation/Emission370 nm, HBW 12 nm
Unit SizeEach
1423802Full specifications
Each380 nm
1,234.00
Your Price:
***
Product TypeExcitation and Emission Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength380 nm
Excitation/Emission380 nm, HBW 11 nm
Unit SizeEach
1423902Full specifications
Each390 nm
672.00
Your Price:
***
Product TypeExcitation Filter
For Use With (Equipment)Fluoroskan™, Fluoroskan™ FL
Wavelength390 nm
Excitation/Emission390 nm, HBW 20 nm
Unit SizeEach
Showing 10 of 67Show more
  • Filters play an essential role in fluorometric measurements
  • When ordering special filters please provide as much information as possible for the best measurement results
  • If advice is needed which filters to order, contact a local Thermo Fisher Scientific representative with the following details:
    • Name of the application
    • Excitation and emission spectra
    • Peak or center wavelengths of the excitation and emission spectra
  • Fluorometric filters can also be used for luminometry depending on the application
  • For luminometric applications requiring filters, contact technical support for instructions

Figures