Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers
Thermo Scientific™

Filters and Accessories for Thermo Scientific Fluoroskan™, Fluoroskan™ FL and Luminoskan™ Microplate Readers

Fluoroskan™, Fluoroskan™FL, and Luminoskan™ microplate fluorometers and luminometers can be equipped with a wide range of accessories, additional excitation filters, and emission filters to expand the instrument's functionality and application flexibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeWavelength
142525LEmission Filter525/30 nm
1423202Excitation Filter320/40 nm
1423352Excitation Filter335/13 nm
142370212Excitation and Emission Filter370/12 nm
1423902Excitation Filter390/20 nm
1424002Excitation and Emission Filter400/10 nm
1424502Excitation and Emission Filter450/11 nm
1424602Excitation and Emission Filter460/24 nm
1424802Excitation and Emission Filter480/10 nm
1424852Excitation and Emission Filter485/16 nm
1424922Excitation Filter492/13 nm
1425102Excitation and Emission Filter510/10 nm
1425172Excitation and Emission Filter517/15 nm
1425182Emission Filter518/13 nm
1425202Excitation and Emission Filter520/10 nm
1425302Excitation Filter530/10 nm
1425382Emission Filter538/25 nm
142540210Excitation Filter540/10 nm
1425442Excitation and Emission Filter544/15 nm
1425552Emission Filter555/25 nm
142560210Excitation and Emission Filter560/10 nm
1425902Emission Filter590/14 nm
1426032Excitation Filter603/7 nm
1426202Excitation and Emission Filter620/10 nm
1426402Excitation and Emission Filter640/10 nm
142640LEmission Filter Long Pass640 nm
1426462Excitation Filter646/10 nm
1426702Excitation and Emission Filter670/10 nm
1426802Excitation and Emission Filter680/20 nm
Catalog number 142525L
Price (USD)
1,354.00
Each
Product Type:
Emission Filter
Wavelength:
525/30 nm
Price (USD)
1,354.00
Each
For running fluorometric or luminometric assays, the Fluoroskan, Fluoroskan FL, and Luminoskan microplate readers offer reliability and optical technology that has already been proven in thousands of laboratories all over the world. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility. Additionally, the Thermo Scientific™ SkanIt™ Software increases efficiency in your lab.
  • Filters play an essential role in fluorometric measurements
  • When ordering special filters please provide as much information as possible for the best measurement results
  • If advice is needed which filters to order, contact a local Thermo Fisher Scientific representative with the following details:
    • Name of the application
    • Excitation and emission spectra
    • Peak or center wavelengths of the excitation and emission spectra
  • Fluorometric filters can also be used for luminometry depending on the application
  • For luminometric applications requiring filters, contact technical support for instructions
Specifications
Excitation/Emission525 nm, HBW 30 nm
Wavelength525/30 nm
For Use With (Equipment)Luminoskan, Fluoroskan FL
Product TypeEmission Filter
Unit SizeEach