Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Thermo Scientific™
Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeWavelength
1425605Optical Filter560 nm
1423405Optical Filter340 nm
1423755Optical Filter375 nm
1424055Optical Filter405 nm
1424145Optical Filter414 nm
1424505Optical Filter450 nm
1424925Optical Filter492 nm
1425205Optical Filter520 nm
1425305Optical Filter530 nm
1425405Optical Filter540 nm
1425505Optical Filter550 nm
1425705Optical Filter570 nm
1425955Optical Filter595 nm
1426205Optical Filter620 nm
1426305Optical Filter630 nm
1426505Optical Filter650 nm
1426905Optical Filter690 nm
1427405Optical Filter740 nm
1427505Optical Filter750 nm
1428205Optical Filter820 nm
Catalog number 1425605
Price (MXN)
-
Product Type:
Optical Filter
Wavelength:
560 nm
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Product LineMultiskan
Wavelength560 nm
For Use With (Equipment)Multiskan FC
Product TypeOptical Filter
Unit SizeEach