DPBS, calcium, magnesium, glucose, pyruvate
Gibco™

DPBS, calcium, magnesium, glucose, pyruvate

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
14287080500 mL
142870721000 mL
產品號碼 14287080
價格 (TWD)
3,640.00
線上優惠
Ends: 31-Dec-2026
4,543.00
您節省 903.00 (20%)
Each
Quantity:
500 mL
Customize this product
價格 (TWD)
3,640.00
線上優惠
Ends: 31-Dec-2026
4,543.00
您節省 903.00 (20%)
Each

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.


This DPBS is modified as follows:
WithWithout
• Calcium• Phenol Red
• Magnesium 
• Glucose 
• Sodium Pyruvate 


The complete formulation is available.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

For Research Use Only. Not for use in diagnostic procedures.
規格
Chemical Name or MaterialDPBS
IncludesSodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Recommended StorageStorage conditions: 2-8° C
Shelf life: 36 months from date of manufacture
SterilitySterile-filtered
Concentration1 X
For Use With (Application)Mammalian Cell Culture
Physical FormLiquid
Product LineGibco
Quantity500 mL
Solution TypeDulbecco's Phosphate Buffered Saline
Unit SizeEach

常見問答集 (常見問題)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.