Dynabeads™ Co-Immunoprecipitation Kit
Invitrogen™

Dynabeads™ Co-Immunoprecipitation Kit

The Dynabeads™ Co-Immunoprecipitation Kit is intended for the coupling of an antibody to the surface of Dynabeads™ M-270 Epoxy beads.Read more
Have Questions?
Catalog NumberQuantity
14321D40 reactions
Catalog number 14321D
Price (USD)
1,052.65
Online exclusive
1,108.00
Save 55.35
Each
Quantity:
40 reactions
Price (USD)
1,052.65
Online exclusive
1,108.00
Save 55.35
Each

The Dynabeads™ Co-Immunoprecipitation Kit is intended for the coupling of an antibody to the surface of Dynabeads™ M-270 Epoxy beads. The coupled beads and supplied buffers can then be used for co-immunoprecipitation (co-IP) of proteins, intact protein complexes, or intact protein-nucleic acid complexes. In addition to epoxy-activated Dynabeads magnetic beads, the kit includes the buffers needed for efficient antibody coupling, gentle washing, and elution of the co-isolated protein complexes.

Benefits of Dynabeads Co-Immunoprecipitation Kit include:
• Convenience—contains optimized buffers and epoxy-activated Dynabeads magnetic beads for efficient co-IP
• Stabile—gentle separation technology preserves native protein conformations and large protein complexes
• Flexibility—allows coupling of any antibody of your choice
• Robustness—covalent antibody coupling to the Dynabeads magnetic beads avoids co-elution of the antibody with the target proteins

Efficient and easy coupling
With this kit, antibodies are covalently coupled to Dynabeads M-270 Epoxy beads, minimizing the risk of bound antibodies contaminating the final eluate. Dynabeads M-270 Epoxy beads exhibit ultra-low background binding and do not require blocking before use. The supplied buffers are optimized to give consistent and reliable results. The kit includes eight different binding, washing, and elution buffers, in addition to Dynabeads M-270 Epoxy beads, to enable purification of complex protein structures of any size. All kit components are analytical grade and compatible with protease and phosphatase inhibitors. Target specific antibody is required for coupling, but not included. The protocol includes coupling, sample preparation (detergent and cryolysis methods), co-IP, and elution and is compatible with western blot/silver stain, Coomassie stain, mass spectrometry, and extraction of nucleic acids from isolated DNA/RNA binding protein complexes.

Magnetic bead–based separation offers easy handling
Captured proteins and protein complexes are easily separated, washed, and eluted using a DynaMag magnet due to the magnetic properties of the Dynabeads M-270 Epoxy beads. The kit is designed for and tested with cultured yeast and mammalian cells and is compatible with tissue, insect, bacterial, and other lysates.

Automation-ready for use with KingFisher purification systems
The 2.8-μm magnetic beads are ideal for high-throughput enrichment. This process can be automated using any of our KingFisher sample purification systems

Commercial supply
Our manufacturing sites are ISO 13485–certified. Bead characteristics and manufacturing conditions help ensure consistency and batch reproducibility, making them an ideal choice for commercial supply. If you are in the process of customizing the Dynabeads Co-Immunoprecipitation Kit in a commercial product/service or need a larger bulk volume of this product, please contact us at oemdynal@lifetech.com or read more on our Dynabeads OEM page

Learn more about Dynabeads magnetic beads and KingFisher Sample Purification Systems:
• Dynabeads M-270 Epoxy beads supplied in the kit can be purchased separately
• See our immunoprecipitation selection table, data, and references
• Dynabeads magnetic beads selection guide
• See magnets for Dynabeads magnetic beads separations
• KingFisher automation protocols
• Watch a video about the KingFisher Flex instrument

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
Final Product TypeProtein Complexes (Intact), Protein/Nucleic Acid Complexes
For Use With (Application)Co-immunoprecipitation
For Use With (Equipment)KingFisher Sample Purification System, DynaMag magnets
FormatKit
High-throughput CompatibilityHigh-throughput Compatible
MaterialMagnetic Polystyrene
Quantity40 reactions
Regulatory StatusRUO
Sample TypeCell Cultures, Disks, Plant, Tissue Digests, Cell Extracts, Cell Lysates, Intact Proteins, Plant, Normalized Buccal Cells
Shelf Life36 months from date of manufacture
Shipping ConditionAmbient Temperature
Sufficient For40 Tests
Surface FunctionalityEpoxy
Product LineDynabeads
TypeCo-IP Kit
Unit SizeEach
Contents & Storage
• Dynabeads M-270 Epoxy (>60 mg freeze-dried)
• Buffer C1 (20 mL)
• Buffer C2 (8 mL)
• Buffer HB (15 mL)
• Buffer LB (15 mL)
• Buffer SB (40 mL)
• 5X IP Buffer (120 mL)
• 5X LWB (24 mL)
• Buffer EB (4 mL)

Store at 2°C to 8°C or room temperature.

Frequently asked questions (FAQs)

My Dynabeads magnetic beads are not pelleting well with the magnet. Do you have any suggestions for me?

Please review the following possibilities for why your Dynabeads magnetic beads are not pelleting:

- The solution is too viscous.
- The beads have formed aggregates because of protein-protein interaction.

Try these suggestions: - Increase separation time (leave tub on magnet for 2-5 minutes)
- Add DNase I to the lysate (~0.01 mg/mL)
- Increase the Tween 20 concentration to ~0.05% of the binding and/or washing buffer.
- Add up to 20 mM beta-merecaptoethanol to the binding and/or wash buffers.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

I have a long double-stranded DNA fragment I would like to isolate. What product do you recommend?

For biotin-labled DNA that is less than 1 kb, we recommend you use Dynabeads M270 Streptavidin and MyOne C1 magnetic beads. We recommend our Dynabeads KilobaseBINDER Kit, which is designed to immobilize long (>1 kb) double-stranded DNA molecules. The KilobaseBINDER reagent consists of M-280 Streptavidin-coupled Dynabeads magnetic beads along with a patented immobilization activator in the binding solution to bind to long, biotinylated DNA molecules for isolation. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/immobilisation-of-long-biotinylated-dna-fragments.html) for more information in regards to long biotinylated DNA fragment isolation.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

Can I use Dynabeads magnetic beads to isolate single-stranded DNA templates?

Yes, Dynabeads magnetic beads can be used to isolate single-stranded DNA. Streptavidin Dynabeads magnetic beads can be used to target biotinylated DNA fragments, followed by denaturation of the double-stranded DNA and removal of the non-biotinylated strand. The streptavidin-coupled Dynabeads magnetic beads will not inhibit any enzymatic activity. This enables further handling and manipulation of the bead-bound DNA directly on the solid phase. Please see the following link (https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/napamisc/capture-of-biotinylated-targets/preparing-single-stranded-dna-templates.html) for more information in regards to single-stranded DNA capture.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

What is the magnetic susceptibility for Dynabeads magnetic beads?

Magnetic susceptibility is a measure of how quickly the beads will migrate to the magnet. This will depend on the iron content and the character of the iron oxide. The magnetic susceptibility given for the Dynabeads magnetic beads is the mass susceptibility, given either as cgs units/g or m^3/kg (the latter being an SI unit). For ferri- and ferromagnetic substances, the magnetic mass susceptibility is dependent upon the magnetic field strength (H), as the magnetization of such substances is not a linear function of H but approaches a saturation value with increasing field. For that reason, the magnetic mass susceptibility of the Dynabeads magnetic beads is determined by a standardized procedure under fixed conditions. The magnetic mass susceptibility given in our catalog is thus the SI unit. Conversion from Gaussian (cgs, emu) units into SI units for magnetic mass susceptibility is achieved by multiplying the Gaussian factor (emu/g or cgs/g) by 4 pi x 10^-3. The resulting unit is also called the rationalized magnetic mass susceptibility, which should be distinguished from the (SI) dimensionless magnetic susceptibility unit. In general, magnetic mass susceptibility is a measure of the force (Fz) influencing an object positioned in a nonhomogenous magnetic field. The magnetic mass susceptibility of the Dynabeads magnetic beads is measured by weighing a sample, and then subjecting the sample to a magnetic field of known strength. The weight (F1) is then measured, and compared to the weight of the sample when the magnetic field is turned off (F0). The susceptibility is then calculated as K x 10^-3 = [(F1-F0) x m x 0.335 x 10^6], where K is the mass susceptibility of the sample of mass m. The susceptibility is then converted to SI units.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.

How can I determine coupling efficiency of Dynabeads magnetic beads?

There are different methods to check binding of ligands to the beads, including optical density (OD) measurement, fluorescent labeling, and radioactive labeling.

For OD measurement, you would measure the OD of the ligand before immobilization to the beads and compare it with the ligand concentration that is left in the supernatant after coating. This gives a crude measurement of how much protein has bound to the beads.

Protocol:

1.Set spectrophotometer to the right wavelength. As a blank, use the Coupling Buffer.
2.Measure the absorbance of the Pre-Coupling Solution. A further dilution may be necessary to read the absorbance, depending upon the amount of ligand added.
3.Measure the absorbance of the Post-Coupling Solution. A dilution may be necessary to read the absorbance.
4.Calculate the coupling efficiency, expressed as the % protein uptake, as follows. [(Pre-Coupling Solution x D) - (Post-Coupling Solution x D)] x 100/(Pre-Coupling Solution x D) where D = dilution factor.

For fluorescent labeling, we suggest negatively quantifying the amount of ligand bound by measuring ligand remaining in the coupling supernatant (compared to the original sample), rather than directly measuring the ligands on the beads. Add labeled ligand to the beads, and measure how much ligand is left in the supernatant (not bound to the beads). By comparing this with the total amount added in the first place, you can then calculate how much of the ligand that has been bound to the beads. Keep in mind that the Dynabeads magnetic beads are also autofluorescent, which is why direct measuring of fluorescence of the bead-bound ligands is not recommended, but rather this indirect approach. The label could be, for example, FITC/PE. Some researchers perform a direct approach with success (using a flow cytometer).

Radioactive labeling is the most sensitive method of the three, but it is also the most difficult one. It involves radioactively labeling a portion of the ligand. We use radiolabeled I-125 in tracer amounts and mix it with "cold" ligands in a known ratio before coupling. The absolute quantities for the ligand on the beads should be obtained by measuring the beads in a scintillation (gamma) counter and comparing the cpm with a standard.

Protocol:

1.Take out an appropriate amount of beads and wash the beads in 1 mL of binding buffer.
2.Pipette out desired amount of human IgG in a separate tube.
3.Mix the human IgG with I-125-labeled human IgG (30,000 - 100,000 cpm).
4.Dilute the mixture of human IgG and I-125-labeled human IgG to 100 mL in binding buffer.
5.Incubate for 30 minutes at room temperature and measure the cpm in a scintillation counter.
6.Wash the beads (with coating) four times, and measure cpm again.
The % binding is calculated by using the equation : (cpm after washing/cpm before washing)x100%.

Find additional tips, troubleshooting help, and resources within our Dynabeads Nucleic Acid Purification Support Center.