Thermo Scientific™

ImmunoCAP™ Grass Pollen Allergens

Catalog number: 14413001
Thermo Scientific™

ImmunoCAP™ Grass Pollen Allergens

Catalog number: 14413001
Quantify specific IgE antibodies with ImmunoCAP™ Grass Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
 
Catalog Number
14413001
Unit Size
Each
Common Name
Sweet vernal grass
Scientific Name
Anthoxanthum odoratum
Code
g1
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameAnthoxanthum odoratum
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameSweet vernal grass
Quantity16 Tests
Codeg1
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Rye-grass
Scientific Name
Lolium perenne
Code
g5
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameLolium perenne
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameRye-grass
Quantity16 Tests
Codeg5
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Meadow grass, Kentucky blue
Scientific Name
Poa pratensis
Code
g8
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NamePoa pratensis
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameMeadow grass, Kentucky blue
Quantity16 Tests
Codeg8
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Timothy grass
Scientific Name
Phleum pratense
Code
g6
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NamePhleum pratense
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameTimothy grass
Quantity16 Tests
Codeg6
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Cultivated rye
Scientific Name
Secale cereale
Code
g12
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameSecale cereale
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameCultivated rye
Quantity16 Tests
Codeg12
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Cocksfoot
Scientific Name
Dactylis glomerata
Code
g3
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameDactylis glomerata
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameCocksfoot
Quantity16 Tests
Codeg3
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Meadow fescue
Scientific Name
Festuca elatior
Code
g4
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameFestuca elatior
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameMeadow fescue
Quantity16 Tests
Codeg4
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Bermuda grass
Scientific Name
Cynodon dactylon
Code
g2
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameCynodon dactylon
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameBermuda grass
Quantity16 Tests
Codeg2
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Velvet grass
Scientific Name
Holcus lanatus
Code
g13
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameHolcus lanatus
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameVelvet grass
Quantity16 Tests
Codeg13
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Johnson grass
Scientific Name
Sorghum halepense
Code
g10
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameSorghum halepense
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameJohnson grass
Quantity16 Tests
Codeg10
Unit SizeEach
Showing 10 of 23Show more
Catalog NumberSpecificationsUnit SizeCommon NameScientific NameCodeQuantityPrice (USD)
14413001Full specifications
EachSweet vernal grassAnthoxanthum odoratumg116 TestsRequest A Quote
Scientific NameAnthoxanthum odoratum
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameSweet vernal grass
Quantity16 Tests
Codeg1
Unit SizeEach
14413401Full specifications
EachRye-grassLolium perenneg516 TestsRequest A Quote
Scientific NameLolium perenne
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameRye-grass
Quantity16 Tests
Codeg5
Unit SizeEach
14413601Full specifications
EachMeadow grass, Kentucky bluePoa pratensisg816 TestsRequest A Quote
Scientific NamePoa pratensis
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameMeadow grass, Kentucky blue
Quantity16 Tests
Codeg8
Unit SizeEach
14410001Full specifications
EachTimothy grassPhleum pratenseg616 TestsRequest A Quote
Scientific NamePhleum pratense
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameTimothy grass
Quantity16 Tests
Codeg6
Unit SizeEach
14410101Full specifications
EachCultivated ryeSecale cerealeg1216 TestsRequest A Quote
Scientific NameSecale cereale
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameCultivated rye
Quantity16 Tests
Codeg12
Unit SizeEach
14411701Full specifications
EachCocksfootDactylis glomeratag316 TestsRequest A Quote
Scientific NameDactylis glomerata
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameCocksfoot
Quantity16 Tests
Codeg3
Unit SizeEach
14413301Full specifications
EachMeadow fescueFestuca elatiorg416 TestsRequest A Quote
Scientific NameFestuca elatior
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameMeadow fescue
Quantity16 Tests
Codeg4
Unit SizeEach
14413101Full specifications
EachBermuda grassCynodon dactylong216 TestsRequest A Quote
Scientific NameCynodon dactylon
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameBermuda grass
Quantity16 Tests
Codeg2
Unit SizeEach
14413901Full specifications
EachVelvet grassHolcus lanatusg1316 TestsRequest A Quote
Scientific NameHolcus lanatus
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameVelvet grass
Quantity16 Tests
Codeg13
Unit SizeEach
14413801Full specifications
EachJohnson grassSorghum halepenseg1016 TestsRequest A Quote
Scientific NameSorghum halepense
Regulatory StatusFDA Cleared
Code TypeGrass
Common NameJohnson grass
Quantity16 Tests
Codeg10
Unit SizeEach
Showing 10 of 23Show more
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing
Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option
Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms
As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text