ImmunoCAP™ Grass Pollen Allergens
Thermo Scientific™

ImmunoCAP™ Grass Pollen Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon Name
1441890116 Testsg17Bahia grass
1441340116 Testsg5Rye-grass
1441360116 Testsg8Meadow grass, Kentucky blue
1441000116 Testsg6Timothy grass
1441010116 Testsg12Cultivated rye
1441170116 Testsg3Cocksfoot
1441300116 Testsg1Sweet vernal grass
1441330116 Testsg4Meadow fescue
1441310116 Testsg2Bermuda grass
1441390116 Testsg13Velvet grass
1441380116 Testsg10Johnson grass
1441370116 Testsg9Redtop, Bentgrass
1441880116 Testsg15Cultivated wheat
1441870116 Testsg7Common reed
1442380116 Testsg11Brome grass
1442400116 Testsg16Meadow foxtail
1442390116 Testsg14Cultivated oat
1443440110 Testsg70Wild rye grass
1443600110 Testsg71Canary grass
1448210110 Testsg201Barley
1448220110 Testsg202Maize, Corn
1451451010 TestsRg203Salt grass
1451541010 TestsRg204False oat-grass
Catalog number 14418901
Price (USD)
-
Quantity:
16 Tests
Code:
g17
Common Name:
Bahia grass
Quantify specific IgE antibodies with ImmunoCAP™ Grass Pollen Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing
Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option
Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms
As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
Codeg17
Common NameBahia grass
Quantity16 Tests
Code TypeGrass
Regulatory StatusFDA Cleared
Scientific NamePaspalum notatum
Unit SizeEach