Thermo Scientific™

ImmunoCAP™ Seed, Legume and Nut Allergens

Catalog number: 14421201
Thermo Scientific™

ImmunoCAP™ Seed, Legume and Nut Allergens

Catalog number: 14421201
Quantify specific IgE antibodies with ImmunoCAP™ Seed, Legume and Nut Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
 
Catalog Number
14421201
Unit Size
Each
Common Name
Coconut
Scientific Name
Cocos nucifera
Code
f36
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameCocos nucifera
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameCoconut
Quantity16 Tests
Codef36
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Soybean
Scientific Name
Glycine max
Code
f14
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameGlycine max
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameSoybean
Quantity16 Tests
Codef14
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Wheat
Scientific Name
Triticum aestivum
Code
f4
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameTriticum aestivum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameWheat
Quantity16 Tests
Codef4
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Peanut
Scientific Name
Arachis hypogaea
Code
f13
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameArachis hypogaea
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NamePeanut
Quantity16 Tests
Codef13
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Buckwheat
Scientific Name
Fagopyrum esculentum
Code
f11
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameFagopyrum esculentum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameBuckwheat
Quantity16 Tests
Codef11
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Maize, Corn
Scientific Name
Zea mays
Code
f8
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameZea mays
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameMaize, Corn
Quantity16 Tests
Codef8
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Rye
Scientific Name
Secale cereale
Code
f5
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameSecale cereale
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameRye
Quantity16 Tests
Codef5
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Barley
Scientific Name
Hordeum vulgare
Code
f6
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameHordeum vulgare
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameBarley
Quantity16 Tests
Codef6
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Oat
Scientific Name
Avena sativa
Code
f7
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NameAvena sativa
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameOat
Quantity16 Tests
Codef7
Unit SizeEach
Catalog Number
Unit Size
Each
Common Name
Pea
Scientific Name
Pisum sativum
Code
f12
Quantity
16 Tests
Price (USD)
Full specifications
Scientific NamePisum sativum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NamePea
Quantity16 Tests
Codef12
Unit SizeEach
Showing 10 of 40Show more
Catalog NumberSpecificationsUnit SizeCommon NameScientific NameCodeQuantityPrice (USD)
14421201Full specifications
EachCoconutCocos nuciferaf3616 TestsRequest A Quote
Scientific NameCocos nucifera
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameCoconut
Quantity16 Tests
Codef36
Unit SizeEach
14411501Full specifications
EachSoybeanGlycine maxf1416 TestsRequest A Quote
Scientific NameGlycine max
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameSoybean
Quantity16 Tests
Codef14
Unit SizeEach
14411301Full specifications
EachWheatTriticum aestivumf416 TestsRequest A Quote
Scientific NameTriticum aestivum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameWheat
Quantity16 Tests
Codef4
Unit SizeEach
14412601Full specifications
EachPeanutArachis hypogaeaf1316 TestsRequest A Quote
Scientific NameArachis hypogaea
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NamePeanut
Quantity16 Tests
Codef13
Unit SizeEach
14412501Full specifications
EachBuckwheatFagopyrum esculentumf1116 TestsRequest A Quote
Scientific NameFagopyrum esculentum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameBuckwheat
Quantity16 Tests
Codef11
Unit SizeEach
14412401Full specifications
EachMaize, CornZea maysf816 TestsRequest A Quote
Scientific NameZea mays
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameMaize, Corn
Quantity16 Tests
Codef8
Unit SizeEach
14412301Full specifications
EachRyeSecale cerealef516 TestsRequest A Quote
Scientific NameSecale cereale
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameRye
Quantity16 Tests
Codef5
Unit SizeEach
14417201Full specifications
EachBarleyHordeum vulgaref616 TestsRequest A Quote
Scientific NameHordeum vulgare
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameBarley
Quantity16 Tests
Codef6
Unit SizeEach
14417301Full specifications
EachOatAvena sativaf716 TestsRequest A Quote
Scientific NameAvena sativa
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NameOat
Quantity16 Tests
Codef7
Unit SizeEach
14417601Full specifications
EachPeaPisum sativumf1216 TestsRequest A Quote
Scientific NamePisum sativum
Regulatory StatusFDA Cleared
Code TypeSeed, Legume, and Nut
Common NamePea
Quantity16 Tests
Codef12
Unit SizeEach
Showing 10 of 40Show more
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text