Thermo Scientific™
Shandon™ Frozen Section Staining Kit
 Related applications:
Anatomical Pathology
Process a limited number of slides in an ideal environment with the Thermo Scientific™ Shandon™ Frozen Section Staining Kit. The kit works exceptionally well for staining tissue mounted on coverglass.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Sturdy cover with piano hinge seals beakers to minimize evaporation
  • Rubber feet on rack protect table surface
  • Glass beakers have a capacity of 200mL (6.75 oz.) and are 10cm (4 in.) tall
  • Slide holder will hold ten 76 x 26mm (3 x 1 in.) slides
  • Stainless-steel support rack measures 3.5 x 4.5 x 17.25 in. when rack is closed
Documents
Manuals & protocols