ImmunoCAP™ Mold and Yeast Allergens
ImmunoCAP™ Mold and Yeast Allergens
Thermo Scientific™

ImmunoCAP™ Mold and Yeast Allergens

Measure specifc IgE antibodies using ImmunoCAP™ allergens, an essential tool to aid in the diagnosis of allergic conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityCodeCommon NameRegulatory Status
1452241010 TestsRm211Trichophyton mentagrophytes var. interdigitaleASR
1441060116 Testsm6Alternaria alternataFDA Cleared
1441050116 Testsm2Cladosporium herbarumFDA Cleared
1441190116 Testsm3Aspergillus fumigatusFDA Cleared
1441200116 Testsm5Candida albicansFDA Cleared
1441450116 Testsm4Mucor racemosusFDA Cleared
1441600116 Testsm1Penicillium chrysogenum (P. notatum)FDA Cleared
1442580116 Testsm7Botrytis cinereaFDA Cleared
1442600116 Testsm9Fusarium proliferatum (F. moniliforme)FDA Cleared
1442590116 Testsm8Setomelanomma rostrata (Helminthosporium halodes)FDA Cleared
1442610116 Testsm10Stemphylium herbarum (S. botryosum)FDA Cleared
1442620116 Testsm11Rhizopus nigricansFDA Cleared
1442640116 Testsm13Phoma betaeFDA Cleared
1442630116 Testsm12Aureobasidium pullulansFDA Cleared
1442660116 Testsm15Trichoderma virideFDA Cleared
1442650116 Testsm14Epicoccum purpurascensFDA Cleared
1442670116 Testsm16Curvularia lunataFDA Cleared
1443490110 Testsm70Pityrosporum orbiculare (syn. Malassezia)FDA Cleared
1444830116 Testsm205Trichophyton rubrumFDA Cleared
1448300110 Testsm202Acremonium kilienseFDA Cleared
1448310110 Testsm207Aspergillus nigerFDA Cleared
1448890110 Testsm80Staphylococcal enterotoxin AFDA Cleared
1448900110 Testsm81Staphylococcal enterotoxin BFDA Cleared
1449401010 TestsRm46Bipolaris spiciferaASR
1450271010 TestsRm203Trichosporon pullulansASR
1450291010 TestsRm201Tilletia triticiASR
1450281010 TestsRm204Ulocladium chartarumASR
1452021010 TestsRm208Chaetomium globosumASR
1452031010 TestsRm209Penicillium glabrumASR
1452231010 TestsRm210Trichophyton mentagrophytes var. goetziiASR
1453011010 TestsRm226Staphylococcal enterotoxin TSSTASR
Catalog number 14522410
Price (USD)
-
Request A Quote
Quantity:
10 Tests
Code:
Rm211
Common Name:
Trichophyton mentagrophytes var. interdigitale
Regulatory Status:
ASR
Quantify specific IgE antibodies with ImmunoCAP™ Mold and Yeast Allergens, essential tools to aid in the diagnosis of allergic conditions. IgE antibodies appear in human serum and plasma as a result of sensitization to a specific allergen. Measurement of circulating IgE antibodies provides an objective assessment of sensitization to an allergen. In general, low IgE antibody levels indicate a low probability of allergic disease, whereas high antibody levels to an allergen show a higher correlation with an allergic disease.
ImmunoCAP Specific IgE offers the laboratory:
  • Truly quantitative measuring range from 0.1 to 100 kUA/L
  • Intra-assay CV (%) comparable to routine clinical chemistry immunoassays
  • Excellent consistency over time, and between countries, systems, labs and persons
Expected test values
ImmunoCAP Specific IgE detects IgE antibodies in the range 0.1 to 100 kUA /l, where A represents allergen-specific antibodies. The result is reported quantitatively. In clinical practice, 0.35 kUA /l has commonly been used as a cut-off. A large number of studies have been performed to evaluate the clinical performance of ImmunoCAP Specific IgE tests in allergy diagnosis. Clinical performance is expressed as sensitivity, ranging from 84 to 95%, and specificity, ranging from 85 to 94%. Sensitivity and specificity were reported from multi-center studies including several hundred patients tested for a range of different allergens.
  • Specimen collection and preparation
  • Use serum and plasma (EDTA or heparin) samples from venous or capillary blood
  • Collect blood samples using standard procedures
  • Keep specimens at room temperature (RT) for shipping purposes only
  • Store at 2–8°C up to one week, otherwise store at –20°C
  • Avoid repeated freezing and thawing

Note: Blood samples for testing ImmunoCAP drugs and venom should be collected during or close to the event, preferably not later than 6 months after exposure. If the test result is negative and an IgE-mediated reaction is still strongly suspected, it is advisable to draw a new sample and repeat the test at 5 to 6 weeks.

The presence of specific IgE antibodies is useful to identify the allergens that elicit symptoms and signs of allergy in patients with respiratory allergic diseases including asthma, food allergy, and anaphylactic sensitivity.

Knowing IgE antibody levels provides guidance to clinicians to:
  • Identify offending allergens
  • Tailor advice to individual patients
  • Indicate development of tolerance (food allergy, specific immunotherapy)
  • Inform optimized individual medical treatment plans
  • Refer to specialists appropriately
  • Evaluate whether or not specific immunotherapy is an option

Specific IgE testing also aids in the identification of patients at risk of:
  • The allergy march – a progression of skin symptoms to respiratory symptoms
  • Severe symptoms including asthma exacerbations – a progression of mild symptoms to severe symptoms
  • Progression of recurrent symptoms to persistent symptoms

As in all diagnostic testing, a definitive clinical diagnosis should be made by the clinician after evaluation of all clinical and laboratory findings.
Specifications
CodeRm211
Common NameTrichophyton mentagrophytes var. interdigitale
Quantity10 Tests
Code TypeMold/Yeast
Regulatory StatusASR
Unit SizeEach