4,4'-Dimethoxytrityl Chloride, 98%
4,4'-Dimethoxytrityl Chloride, 98%
4,4'-Dimethoxytrityl Chloride, 98%
Thermo Scientific Chemicals

4,4'-Dimethoxytrityl Chloride, 98%

CAS: 40615-36-9 | C21H19ClO2 | 338.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 146021000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS40615-36-9
IUPAC Name1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
Molecular FormulaC21H19ClO2
InChI KeyJBWYRBLDOOOJEU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric97.5 to 102.5 % (ex Cl)
Infrared spectrumConforms
Appearance (Form)Powder
Melting point121°C to 127°C
Appearance (Color)Pink to orange
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only