Adamantano, + 99 %, Thermo Scientific Chemicals
Adamantano, + 99 %, Thermo Scientific Chemicals
Adamantano, + 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Adamantano, + 99 %, Thermo Scientific Chemicals

CAS: 281-23-2 | C10H16 | 136.24 g/mol
Have Questions?
Número de catálogoCantidadEnvase
146111000100 gFrasco de vidrio
Número de catálogo 146111000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS281-23-2
IUPAC Nameadamantane
Molecular FormulaC10H16
InChI KeyORILYTVJVMAKLC-UHFFFAOYSA-N
SMILESC1C2CC3CC1CC(C2)C3
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Appearance (Color)White to beige
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only