Thermo Scientific™

3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific™

Each
Thermo Scientific™

3-Aminoquinuclidine dihydrochloride, 98%, Thermo Scientific™

Each

Chemical Identifiers

CAS
6530-09-2
IUPAC Name
dihydrogen 1-azabicyclo[2.2.2]octan-3-amine dichloride
Molecular Formula
C7H16Cl2N2
InChI Key
STZHBULOYDCZET-UHFFFAOYNA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC1CN2CCC1CC2
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures