2-Bromo-2'-methoxyacetophenone, 98%
2-Bromo-2'-methoxyacetophenone, 98%
2-Bromo-2'-methoxyacetophenone, 98%
Thermo Scientific Chemicals

2-Bromo-2'-methoxyacetophenone, 98%

CAS: 31949-21-0 | C9H9BrO2 | 229.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 146270010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS31949-21-0
IUPAC Name2-bromo-1-(2-methoxyphenyl)ethan-1-one
Molecular FormulaC9H9BrO2
InChI KeyGKNCPTLOPRDYMH-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)White to brown
Appearance (Form)Crystalline powder or solid
Infrared spectrumConforms
Melting point42°C to 47°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only