4-Bromoaniline, 99+%
4-Bromoaniline, 99+%
4-Bromoaniline, 99+%
Thermo Scientific Chemicals

4-Bromoaniline, 99+%

CAS: 106-40-1 | C6H6BrN | 172.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 146280250
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
102.65
Online exclusive
114.00
Save 11.35
Each
Chemical Identifiers
CAS106-40-1
IUPAC Name4-bromoaniline
Molecular FormulaC6H6BrN
InChI KeyWDFQBORIUYODSI-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Appearance (Color)Off-white to grey-brown
Appearance (Form)Crystalline powder
Melting point60°C to 64°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only