4-Bromoaniline, 99+%
4-Bromoaniline, 99+%
4-Bromoaniline, 99+%
Acros Organics

4-Bromoaniline, 99+%

CAS: 106-40-1 | C6H6BrN | 172.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 146281000
HSN 29214290
Price (INR)
43,139.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
43,139.00
Each
Chemical Identifiers
CAS106-40-1
IUPAC Name4-bromoaniline
Molecular FormulaC6H6BrN
InChI KeyWDFQBORIUYODSI-UHFFFAOYSA-N
SMILESNC1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=99.0 %
Melting point60°C to 64°C
Appearance (Color)Off-white to grey-brown
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only