1-(2-Piridilazo)-2-naftol, 98 %, puro, grado indicador
1-(2-Piridilazo)-2-naftol, 98 %, puro, grado indicador
1-(2-Piridilazo)-2-naftol, 98 %, puro, grado indicador
Thermo Scientific Chemicals

1-(2-Piridilazo)-2-naftol, 98 %, puro, grado indicador

CAS: 85-85-8 | C15H11N3O | 249.27 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
10 g
50 g
Envase:
Frasco de vidrio
Número de catálogo 146310050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS85-85-8
IUPAC Name(1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular FormulaC15H11N3O
InChI KeyRAXUMGMWXZYADR-SDXDJHTJSA-N
SMILESO=C1C=CC2=CC=CC=C2C1=NNC1=CC=CC=N1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Suitability(as complexom. indicator) Passes test
Appearance (Color)Orange to red-brown to brown
Titration with HClO4>=97.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicador
RUO – Research Use Only