1-Chloro-4-iodobenzene, 99%
1-Chloro-4-iodobenzene, 99%
1-Chloro-4-iodobenzene, 99%
Thermo Scientific Chemicals

1-Chloro-4-iodobenzene, 99%

CAS: 637-87-6 | C6H4ClI | 238.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 146481000
Price (EUR)
113,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
113,00
Each
Chemical Identifiers
CAS637-87-6
IUPAC Name1-chloro-4-iodobenzene
Molecular FormulaC6H4ClI
InChI KeyGWQSENYKCGJTRI-UHFFFAOYSA-N
SMILESClC1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to light beige
GC>=98.5 %
Infrared spectrumConforms
Melting point52°C to 55°C
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only