Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Thermo Scientific Chemicals

Safranin O, 95%, pure, high purity biological stain

Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation. | CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol
製品番号(カタログ番号)数量パッケージ
14664025025 gGlass bottle
製品番号(カタログ番号) 146640250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS477-73-6
IUPAC Name2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular FormulaC20H19ClN4
InChI KeyQRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
さらに表示
Infrared spectrumConforms
Loss on drying=<10 %
Appearance (Form)Powder
Appearance (Color)Green to red-brown
Dye content>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only