Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Safranin O, 95%, pure, high purity biological stain
Thermo Scientific Chemicals

Safranin O, 95%, pure, high purity biological stain

Safranine O, C20H19ClN4, CAS Number-477-73-6, 3, 7-Diamino-2, 8-dimethyl-5-phenylphenazinium chloride, Basic Red 2, C.I. 50240, 100g, 95%, Brown or Brown-Red to Green, 94% min., 350.84, Pure, Warning, GHS H Statement: Causes serious eye irritation. Causes skin irritation. | CAS: 477-73-6 | C20H19ClN4 | 350.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 146641000
Price (USD)
142.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
142.00
Each
Chemical Identifiers
CAS477-73-6
IUPAC Name2,7-diamino-1,8-dimethyl-5-phenyl-5λ⁵-phenazin-5-ylium chloride
Molecular FormulaC20H19ClN4
InChI KeyQRYAEWIQIBAZOJ-UHFFFAOYSA-N
SMILES[Cl-].CC1=C(N)C=C2C(=C1)N=C1C(C)=C(N)C=CC1=[N+]2C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 %
Appearance (Color)Green to red-brown
Dye content>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only