2-Hydroxybenzophenone, 99%
2-Hydroxybenzophenone, 99%
2-Hydroxybenzophenone, 99%
Thermo Scientific Chemicals

2-Hydroxybenzophenone, 99%

CAS: 117-99-7 | C13H10O2 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 146920100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS117-99-7
IUPAC Name2-benzoylphenol
Molecular FormulaC13H10O2
InChI KeyHJIAMFHSAAEUKR-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Melting point37°C to 40°C
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only