Pentafluorobenzenesulfonyl chloride, 99%
Pentafluorobenzenesulfonyl chloride, 99%
Pentafluorobenzenesulfonyl chloride, 99%
Thermo Scientific Chemicals

Pentafluorobenzenesulfonyl chloride, 99%

CAS: 832-53-1 | C6ClF5O2S | 266.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 147110050
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.50
Save 12.85
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.50
Save 12.85
Each
Chemical Identifiers
CAS832-53-1
IUPAC Namepentafluorobenzene-1-sulfonyl chloride
Molecular FormulaC6ClF5O2S
InChI KeyUOJCTEGNHXRPKO-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(C(F)=C1F)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Infrared spectrumConforms
Refractive index1.4780 to 1.4800 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only