Piperidine, 99%, extra pure
Thermo Scientific Chemicals

Piperidine, 99%, extra pure

CAS: 110-89-4 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
10 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 147180025
Price (EUR)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS110-89-4
IUPAC Namepiperidine
Molecular FormulaC5H11N
InChI KeyNQRYJNQNLNOLGT-UHFFFAOYSA-N
SMILESC1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
Color scale=<50 APHA
Refractive index1.4510 to 1.4540 (20°C, 589 nm)
Water=<0.5 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only