meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals
meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals
meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

meso-2,3-Dibromosuccinic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
147575000500 gPlastic bottle
Catalog number 147575000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS608-36-6
IUPAC Name(2R,3S)-2,3-dibromobutanedioate
Molecular FormulaC4H2Br2O4
InChI KeyFJWGRXKOBIVTFA-XIXRPRMCSA-L
SMILES[O-]C(=O)[C@@H](Br)[C@@H](Br)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Infrared spectrumConforms
Appearance (Form)Powder
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only