p-Toluenesulfonamide, 99%
p-Toluenesulfonamide, 99%
p-Toluenesulfonamide, 99%
Thermo Scientific Chemicals

p-Toluenesulfonamide, 99%

CAS: 70-55-3 | C7H9NO2S | 171.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
產品號碼 147690010
價格 (TWD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS70-55-3
IUPAC Name4-methylbenzene-1-sulfonamide
Molecular FormulaC7H9NO2S
InChI KeyLMYRWZFENFIFIT-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)S(N)(=O)=O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point136°C to 140°C
HPLC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only