2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
2,4-Dichlorophenol, 99%
Thermo Scientific Chemicals

2,4-Dichlorophenol, 99%

CAS: 120-83-2 | C6H4Cl2O | 163.00 g/mol
数量:
5 g
100 g
500 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 147720050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-83-2
IUPAC Name2,4-dichlorophenol
Molecular FormulaC6H4Cl2O
InChI KeyHFZWRUODUSTPEG-UHFFFAOYSA-N
SMILESOC1=CC=C(Cl)C=C1Cl
さらに表示
Appearance (Color)White to beige
Appearance (Form)Crystalline solid
Infrared spectrumConforms
Melting point41°C to 44°C
GC>=98.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only