Nicotinic acid N-oxide, 99%
Nicotinic acid N-oxide, 99%
Nicotinic acid N-oxide, 99%
Thermo Scientific Chemicals

Nicotinic acid N-oxide, 99%

CAS: 2398-81-4 | C6H6NO3 | 140.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 147785000
Price (TWD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2398-81-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only