Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis

Biological buffer, Chelation/Complexation reagent | CAS: 6381-92-6 | C10H18N2Na2O10 | 372.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 147850050
Price (EUR)
676,00
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (EUR)
676,00
Each
Chemical Identifiers
CAS6381-92-6
IUPAC Namedisodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
Molecular FormulaC10H18N2Na2O10
InChI KeyOVBJJZOQPCKUOR-UHFFFAOYSA-L
SMILESO.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Assay99.0 to 101.0 % (On dry substance)
pH4.0 to 6.0 (1 % at 20°C)
Iron (Fe)=<80 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer, Chelation/Complexation reagent
RUO – Research Use Only