2,2-Bis(hydroxymethyl)propionic acid, 98%
2,2-Bis(hydroxymethyl)propionic acid, 98%
2,2-Bis(hydroxymethyl)propionic acid, 98%
Thermo Scientific Chemicals

2,2-Bis(hydroxymethyl)propionic acid, 98%

CAS: 4767-03-7 | C5H10O4 | 134.13 g/mol
数量:
500 g
1 kg
5 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 147970010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4767-03-7
IUPAC Name3-hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
Molecular FormulaC5H10O4
InChI KeyPTBDIHRZYDMNKB-UHFFFAOYSA-N
SMILESCC(CO)(CO)C(O)=O
さらに表示
Appearance (Color)White
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only