2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%
2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%
2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%
Thermo Scientific Chemicals

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride, 97%

CAS: 69812-29-9 | C6H7ClN2O3S2 | 254.71 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1483600101 gGlass bottle
Catalog number 148360010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS69812-29-9
IUPAC Name2-acetamido-4-methyl-1,3-thiazole-5-sulfonyl chloride
Molecular FormulaC6H7ClN2O3S2
InChI KeyNXGKPRKPUCSEIL-UHFFFAOYSA-N
SMILESCC(=O)NC1=NC(C)=C(S1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only