2-Chloroacetamide, 98%
2-Chloroacetamide, 98%
2-Chloroacetamide, 98%
Thermo Scientific Chemicals

2-Chloroacetamide, 98%

CAS: 79-07-2 | C2H4ClNO | 93.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number 148410010
Price (EUR)
84,00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
84,00
Each
Chemical Identifiers
CAS79-07-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
GC>=97.5 %
Ammonium chloride (NH4Cl)=<0.5 %
Water=<0.2 %
Melting point116°C to 120°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only