Benzenesulfonamide, 98+%
Benzenesulfonamide, 98+%
Benzenesulfonamide, 98+%
Thermo Scientific Chemicals

Benzenesulfonamide, 98+%

CAS: 98-10-2 | C6H7NO2S | 157.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 148495000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS98-10-2
IUPAC Namebenzenesulfonamide
Molecular FormulaC6H7NO2S
InChI KeyKHBQMWCZKVMBLN-UHFFFAOYSA-N
SMILESNS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point150°C to 154°C
Appearance (Form)Powder or crystals and/or chunks
Appearance (Color)White to off-white
Infrared spectrumConforms
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only