Ácido 4-bromobenzoico, 97 %, Thermo Scientific Chemicals
Ácido 4-bromobenzoico, 97 %, Thermo Scientific Chemicals
Ácido 4-bromobenzoico, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 4-bromobenzoico, 97 %, Thermo Scientific Chemicals

CAS: 586-76-5 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
500 g
Envase:
Frasco de plástico
Número de catálogo 148701000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS586-76-5
IUPAC Name4-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyTUXYZHVUPGXXQG-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Br)C=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to yellow to beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point251°C to 257°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only