3,3'-Dithiodipropionic acid, 98%
3,3'-Dithiodipropionic acid, 98%
3,3'-Dithiodipropionic acid, 98%
Thermo Scientific Chemicals

3,3'-Dithiodipropionic acid, 98%

CAS: 1119-62-6 | C6H10O4S2 | 210.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 148900500
Price (EUR)
20,65
Online exclusive
22,80
Save 2,15
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
20,65
Online exclusive
22,80
Save 2,15
Each
Chemical Identifiers
CAS1119-62-6
IUPAC Name3-[(2-carboxyethyl)disulfanyl]propanoic acid
Molecular FormulaC6H10O4S2
InChI KeyYCLSOMLVSHPPFV-UHFFFAOYSA-N
SMILESOC(=O)CCSSCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Melting point154°C to 158°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only