Propionic acid, sodium salt, 99.0-100.5%
Propionic acid, sodium salt, 99.0-100.5%
Propionic acid, sodium salt, 99.0-100.5%
Thermo Scientific Chemicals

Propionic acid, sodium salt, 99.0-100.5%

CAS: 137-40-6 | C3H5NaO2 | 96.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 kg
Packaging:
Plastic bottle
Catalog number 149010250
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
25.60
Save 3.95
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
21.65
Special offer
Online exclusive
Ends: 28-Dec-2026
25.60
Save 3.95
Each
Chemical Identifiers
CAS137-40-6
IUPAC Namesodium propanoate
Molecular FormulaC3H5NaO2
InChI KeyJXKPEJDQGNYQSM-UHFFFAOYSA-M
SMILES[Na+].CCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline or granular powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only