1-Bromoadamantane, 99%
1-Bromoadamantane, 99%
1-Bromoadamantane, 99%
Thermo Scientific Chemicals

1-Bromoadamantane, 99%

CAS: 768-90-1 | C10H15Br | 215.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 149031000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS768-90-1
IUPAC Name1-bromoadamantane
Molecular FormulaC10H15Br
InChI KeyVQHPRVYDKRESCL-UHFFFAOYSA-N
SMILESBrC12CC3CC(CC(C3)C1)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige to orange
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point116°C to 120°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only