3-Heptanol, 98%
3-Heptanol, 98%
3-Heptanol, 98%
Thermo Scientific Chemicals

3-Heptanol, 98%

CAS: 589-82-2 | C7H16O | 116.2 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 149090250
価格(JPY)
-
数量:
25 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS589-82-2
IUPAC Nameheptan-3-ol
Molecular FormulaC7H16O
InChI KeyRZKSECIXORKHQS-UHFFFAOYNA-N
SMILESCCCCC(O)CC
さらに表示
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
GC>=97.5 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only