o-Anisaldehyde, 98%
o-Anisaldehyde, 98%
o-Anisaldehyde, 98%
Thermo Scientific Chemicals

o-Anisaldehyde, 98%

CAS: 135-02-4 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 149221000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS135-02-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to light brown
Appearance (Form)Crystals or solid or liquids
Infrared spectrumConforms
Melting point34°C to 40°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only