Isopropanol, 99.5+%, pure
Isopropanol, 99.5+%, pure
Isopropanol, 99.5+%, pure
Thermo Scientific Chemicals

Isopropanol, 99.5+%, pure

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
10 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 149320025
Price (EUR)
81,65
Online Exclusive
91,00
Save 9,35 (10%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
81,65
Online Exclusive
91,00
Save 9,35 (10%)
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Infrared spectrumConforms
GC>=99.50 %
Water=<0.2 % (K.F.)
Residue after evaporation=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only