Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Thermo Scientific Chemicals

Phenol, 99%, extra pure

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 149340051
Price (TWD)
-
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Assay Percent Range98.5% min. (GC)
Beilstein06, 110
CAS108-95-2
ChEBICHEBI:15882
Chemical Name or MaterialPhenol
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only