Phenol, 99%, extra pure
Phenol, 99%, extra pure
Phenol, 99%, extra pure
Thermo Scientific Chemicals

Phenol, 99%, extra pure

CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
1 kg
2.5 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 149340500
Price (USD)
38.80
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
38.80
Each
Chemical Identifiers
CAS108-95-2
IUPAC Namephenol
Molecular FormulaC6H6O
InChI KeyISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point39°C to 42°C
Appearance (Color)Colorless to white
Appearance (Form)Crystals or fused mass
Infrared spectrumConforms
Water=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only