4-Methyl-2-pentanol, 99+%
4-Methyl-2-pentanol, 99+%
4-Methyl-2-pentanol, 99+%
Thermo Scientific Chemicals

4-Methyl-2-pentanol, 99+%

CAS: 108-11-2 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 149380050
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.20
Save 4.55
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
29.65
Special offer
Online exclusive
Ends: 28-Dec-2026
34.20
Save 4.55
Each
Chemical Identifiers
CAS108-11-2
IUPAC Name4-methylpentan-2-ol
Molecular FormulaC6H14O
InChI KeyWVYWICLMDOOCFB-UHFFFAOYNA-N
SMILESCC(C)CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4100 to 1.4120 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only