Acetonitrile, 99+%, extra pure
Acetonitrile, 99+%, extra pure
Acetonitrile, 99+%, extra pure
Thermo Scientific Chemicals

Acetonitrile, 99+%, extra pure

CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number 149520250
Price (EUR)
1.957,00
Each
Quantity:
25 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (EUR)
1.957,00
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.005 %
Color scale=<20 APHA
Water=<0.3 % (K.F.)
Appearance (Form)Clear liquid
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only