Disposable Cuvettes
Fisher Scientific™

Disposable Cuvettes

Choice of polystyrene or methacrylate
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)유형물질Wavelength Range
149551294.5 mLCuvettePolystyrene340 to 750 nm
149551264.5 mLStandard CuvettePMMA285 to 750 nm
14955130
FISHERTOPSELLING-14955130-CS으로도 사용됨
4.5 mLCuvettePMMA285 to 750 nm
14955125
FISHERTOPSELLING-14955125-CS으로도 사용됨
4.5 mLStandard CuvettePolystyrene340 to 750 nm
14955127
FISHERTOPSELLING-14955127-EACH으로도 사용됨
1.5 mLSemi-micro CellPolystyrene340 to 750 nm
14955128
FISHERTOPSELLING-14955128-CS으로도 사용됨
1.5 mLSemi-micro CellMethacrylate285 to 750 nm
카탈로그 번호 14955129
제품 가격(KRW)
-
용량(미터법):
4.5 mL
유형:
Cuvette
물질:
Polystyrene
Wavelength Range:
340 to 750 nm
Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Choice of 4.5 and 1.5mL sample capacity
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

사양
설명Four-clear-sided
일회용Single-use
길이(미터법)12mm
수량500 Case
용량(미터법)4.5 mL
용도(애플리케이션)For assays in visible spectral range, accuracy throughout UV/Visible range
용도 (장비)Spectrophotometers, Fluorometers, Nephelometers
높이(미터법)45 mm
물질Polystyrene
경로 길이10 mm
유형Cuvette
Wavelength Range340 to 750 nm
폭(미터법)12 mm
Unit SizeCase of 500