Acetic acid, potassium salt, 97%, pure, anhydrous
Acetic acid, potassium salt, 97%, pure, anhydrous
Acetic acid, potassium salt, 97%, pure, anhydrous
Thermo Scientific Chemicals

Acetic acid, potassium salt, 97%, pure, anhydrous

CAS: 127-08-2 | C2H3KO2 | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number 149600025
Price (USD)
116.65
Special offer
Online exclusive
Ends: 28-Dec-2026
137.00
Save 20.35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
116.65
Special offer
Online exclusive
Ends: 28-Dec-2026
137.00
Save 20.35
Each
Chemical Identifiers
CAS127-08-2
IUPAC Namepotassium acetate
Molecular FormulaC2H3KO2
InChI KeySCVFZCLFOSHCOH-UHFFFAOYSA-M
SMILES[K+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Appearance (Form)Crystalline or granular powder
Infrared spectrumConforms
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only