Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Acetic acid, sodium salt, 99+%, extra pure, anhydrous
Thermo Scientific Chemicals

Acetic acid, sodium salt, 99+%, extra pure, anhydrous

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
5 kg
Packaging:
View all
Plastic drum
Catalog number 149610050
Price (USD)
306.65
Special offer
Online exclusive
Ends: 28-Dec-2026
360.00
Save 53.35
Each
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (USD)
306.65
Special offer
Online exclusive
Ends: 28-Dec-2026
360.00
Save 53.35
Each
Chemical Identifiers
CAS127-09-3
IUPAC Namesodium acetate
Molecular FormulaC2H3NaO2
InChI KeyVMHLLURERBWHNL-UHFFFAOYSA-M
SMILES[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
Titration with HClO4>=99.0 %
Loss on drying=<1 %
Chloride (Cl)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only